Ertugliflozin 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S,3S,4R,5S)-5-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-1-(hydroxymethyl)-6,8-dioxabicyclo[3.2.1]octane-2,3,4-triol, (1S,2S,3S,4R,5S)-5-(4-Chloro-3-(4-ethoxybenzyl)phenyl)-1-hydroxymethyl-6,8-dioxabicyclo(3.2.1)octane-2,3,4-triol, PF-04971729

A191432-1MG A191432-5MG A191432-100MG A191432-250MG
77151-896EA 6 USD
77151-896 77151-898 77685-458 77685-459
Ertugliflozin 98%
Ertugliflozin

Precaución: for Research Use Only

Formula: C₂₂H₂₅ClO₇
MW: 436.89 g/mol
MDL Number: MFCD21609259
CAS Number: 1210344-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR