5-Fluoropyrimidine-4,6-diol 96%

Supplier: Ambeed
Warning

A191309-1G A191309-5G
77151-892EA 18 USD
77151-892 77151-894
5-Fluoropyrimidine-4,6-diol 96%
5-Fluoropyrimidine-4,6-diol

Precaución: for Research Use Only

Formula: C₄H₃FN₂O₂
MDL Number: MFCD09033260
CAS Number: 106615-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR