Methyl 2-methyl-3-morpholinobenzoate 97%

Supplier: Ambeed
Warning

A191286-1G A191286-250MG A191286-5G
77151-886EA 41 USD
77151-886 77151-888 77151-890
Methyl 2-methyl-3-morpholinobenzoate 97%
Methyl 2-methyl-3-morpholinobenzoate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MDL Number: MFCD06740125
CAS Number: 886497-09-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR