3-Ethynyloxetan-3-ol 98%
Supplier: Ambeed
|
Warning
|
A191260-100MG
A191260-1G
A191260-250MG
A191260-5G
77151-878EA
23
USD
77151-878
77151-880
77151-882
77151-884
3-Ethynyloxetan-3-ol 98%
3-Ethynyloxetan-3-ol
Precaución: for Research Use Only
Formula:
C₅H₆O₂ |
MDL Number:
MFCD20444449 CAS Number: 1352492-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...