1-Benzyl-4-hydroxypiperidine-4-carboxylic acid hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A190795-100MG
A190795-1G
A190795-250MG
A190795-5G
77151-866EA
42.23
USD
77151-866
77151-868
77151-870
77151-872
1-Benzyl-4-hydroxypiperidine-4-carboxylic acid hydrochloride 97%
1-Benzyl-4-hydroxypiperidine-4-carboxylic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₈ClNO₃ MW: 271.74 g/mol |
CAS Number:
1262988-77-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...