(7S,12aS)-7,8-Dihydro-7-(hydroxymethyl)-6H-dibenz[b,d]oxocin-3,7,10,11-tetrol 95%

Supplier: Ambeed
Warning

Synonyms: Protosappanin B

A189663-1MG A189663-5MG
77151-854EA 46.35 USD
77151-854 77151-856
(7S,12aS)-7,8-Dihydro-7-(hydroxymethyl)-6H-dibenz[b,d]oxocin-3,7,10,11-tetrol 95%
(7S,12aS)-7,8-Dihydro-7-(hydroxymethyl)-6H-dibenz[b,d]oxocin-3,7,10,11-tetrol

Precaución: for Research Use Only

Formula: C₁₆H₁₆O₆
MW: 304.29 g/mol
MDL Number: MFCD29048516
CAS Number: 102036-29-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR