9,10-Bis(bromomethyl)anthracene 95%

Supplier: Ambeed
Danger

A189512-1G A189512-100MG A189512-250MG
77151-852EA 194 USD
77151-852 77901-484 77901-717
9,10-Bis(bromomethyl)anthracene 95%
9,10-Bis(bromomethyl)anthracene

Precaución: for Research Use Only

Formula: C₁₆H₁₂Br₂
MDL Number: MFCD00092257
CAS Number: 34373-96-1
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR