H-D-Asp(OBzl)-OBzl·HCl 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Asp(OBzl)-OBzl.HCl

A189475-10G A189475-1G
77151-848EA 51.5 USD
77151-848 77151-850
H-D-Asp(OBzl)-OBzl·HCl 97%
H-D-Asp(OBzl)-OBzl·HCl

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₄·HCl
MW: 349.8 g/mol
MDL Number: MFCD15071452
CAS Number: 174457-99-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR