4-Amino-2-chloronicotinic acid 97%

Supplier: Ambeed
Warning

A189126-100MG A189126-10G A189126-1G A189126-250MG A189126-5G
77151-840EA 46 USD
77151-840 77151-842 77151-844 77151-846 77900-773
4-Amino-2-chloronicotinic acid 97%
4-Amino-2-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD13188881
CAS Number: 1018678-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR