(3-Iodophenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Iodobenzyl cyanide, 2-(3-Iodophenyl)acetonitrile

A188969-1G A188969-250MG A188969-25G
77151-834EA 37.08 USD
77151-834 77151-836 77151-838
(3-Iodophenyl)acetonitrile 98%
(3-Iodophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₆IN
MW: 243.05 g/mol
MDL Number: MFCD00040890
CAS Number: 130723-54-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR