2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one 98%

Supplier: Ambeed
Warning

A188648-100MG A188648-1G A188648-250MG A188648-5G
77151-826EA 28.84 USD
77151-826 77151-828 77151-830 77151-832
2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one 98%
2-Chlorothieno[3,2-d]pyrimidin-4(3H)-one

Precaución: for Research Use Only

Formula: C₆H₃ClN₂OS
MW: 186.619 g/mol
MDL Number: MFCD19382468
CAS Number: 1245811-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR