2-Oxetanemethanamine 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
2-(Aminomethyl)oxetane, Oxetan-2-ylmethanamine
A187399-100MG
A187399-250MG
77151-814EA
88.58
USD
77151-814
77151-816
2-Oxetanemethanamine 95%
2-Oxetanemethanamine
Precaución: for Research Use Only
Formula:
C₄H₉NO MW: 87.12 g/mol |
MDL Number:
MFCD17214171 CAS Number: 882402-12-2 UN: 2735 ADR: 8,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...