4,5-Dichloroindole 95%

Supplier: Ambeed
Warning

A186634-100MG A186634-250MG A186634-1G
77151-810EA 13 USD
77151-810 77151-812 77567-718
4,5-Dichloroindole 95%
4,5-Dichloroindole

Precaución: For research use only.

Formula: C₈H₅Cl₂N
MDL Number: MFCD16610777
CAS Number: 122509-73-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR