5,7-Dibromo-1-indanone 95%

Supplier: Ambeed
Warning

A186126-1G A186126-250MG A186126-5G A186126-25G
77151-792EA 20 USD
77151-792 77151-794 77151-796 77901-016
5,7-Dibromo-1-indanone 95%
5,7-Dibromo-1-indanone

Precaución: for Research Use Only

Formula: C₉H₆Br₂O
MDL Number: MFCD11707100
CAS Number: 923977-18-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR