6-Bromo-4-chloro-7-methoxyquinoline 98%

Supplier: Ambeed
Warning

A185644-100MG A185644-1G A185644-250MG A185644-5G
77151-778EA 23.69 USD
77151-778 77151-780 77151-782 77151-784
6-Bromo-4-chloro-7-methoxyquinoline 98%
6-Bromo-4-chloro-7-methoxyquinoline

Precaución: for Research Use Only

Formula: C₁₀H₇BrClNO
MW: 272.52 g/mol
MDL Number: MFCD22126059
CAS Number: 476660-71-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR