N-Boc-L-isoleucine hemihydrate 96%

Supplier: Ambeed
Warning

Synonyms: Boc-Ile-OH·1/2H2O, Boc-L-isoleucine hemihydrate

A185460-100G A185460-25G A185460-5G
77151-766EA 61 USD
77151-766 77151-768 77151-770 77151-772
N-Boc-L-isoleucine hemihydrate 96%
N-Boc-L-isoleucine hemihydrate

Precaución: for Research Use Only

Formula: C₂₂H₄₄N₂O₉
MW: 480.59 g/mol
MDL Number: MFCD00672705
CAS Number: 204138-23-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR