5-Bromo-1,2-dimethyl-3-(trifluoromethyl)benzene 95%

Supplier: Ambeed
Warning

A185382-1G A185382-250MG A185382-5G
77151-760EA 96.82 USD
77151-760 77151-762 77151-764
5-Bromo-1,2-dimethyl-3-(trifluoromethyl)benzene 95%
5-Bromo-1,2-dimethyl-3-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₉H₈BrF₃
MW: 253.05 g/mol
MDL Number: MFCD29044867
CAS Number: 80245-29-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR