BMS-754807 98+%

Supplier: Ambeed
Warning

Synonyms: BMS754807, (S)-1-(4-((5-cyclopropyl-1H-pyrazol-3-yl)amino)pyrrolo[2,1-f][1,2,4]triazin-2-yl)-N-(6-fluoropyridin-3-yl)-2-methylpyrrolidine-2-carboxamide

A184967-10MG A184967-1MG A184967-25MG A184967-5MG
77151-740EA 139.05 USD
77151-740 77151-742 77151-744 77151-746
BMS-754807 98+%
BMS-754807

Precaución: for Research Use Only

Formula: C₂₃H₂₄FN₉O
MW: 461.49 g/mol
MDL Number: MFCD18633202
CAS Number: 1001350-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR