DL-Tryptophan ethyl ester hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A184739-100G
A184739-25G
A184739-5G
A184739-1G
77151-724EA
367
USD
77151-724
77151-726
77151-728
77901-289
DL-Tryptophan ethyl ester hydrochloride 98%
DL-Tryptophan ethyl ester hydrochloride
Precaución: for Research Use Only
Formula:
C₁₃H₁₆N₂O₂ |
MDL Number:
MFCD00067553 CAS Number: 6519-67-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...