5-Methoxy-1H-indazole-3-carboxylic acid 96%

Supplier: Ambeed
Warning

A184698-100MG A184698-250MG
77151-714EA 32.96 USD
77151-714 77151-716
5-Methoxy-1H-indazole-3-carboxylic acid 96%
5-Methoxy-1H-indazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₈N₂O₃
MW: 192.171 g/mol
MDL Number: MFCD03840712
CAS Number: 90417-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR