GSK-2656157 98%

Supplier: Ambeed
Warning

Synonyms: GSK2656157, 1-(5-(4-Amino-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-5-yl)-4-fluoroindolin-1-yl)-2-(6-methylpyridin-2-yl)ethanone

A184177-1MG A184177-5MG
77151-708EA 31.93 USD
77151-708 77151-710
GSK-2656157 98%
GSK-2656157

Precaución: for Research Use Only

Formula: C₂₃H₂₁FN₆O
MW: 416.46 g/mol
MDL Number: MFCD27997885
CAS Number: 1337532-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR