5-Bromo-2-chloronicotinic acid 97%

Supplier: Ambeed
Warning

A183534-100G A183534-10G A183534-1G A183534-25G A183534-500G A183534-5G
77151-690EA 154.5 USD
77151-690 77151-692 77151-694 77151-696 77151-698 77151-700
5-Bromo-2-chloronicotinic acid 97%
5-Bromo-2-chloronicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃BrClNO₂
MDL Number: MFCD03844847
CAS Number: 29241-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR