1-Methyl-3-oxocyclobutane-1-carboxylic acid 97%

Supplier: Ambeed
Danger

Synonyms: 1-Methyl-3-oxocyclobutanecarboxylic acid

A183424-1G A183424-5G A183424-250MG
77151-688EA 87 USD
77151-688 77593-178 77685-428
1-Methyl-3-oxocyclobutane-1-carboxylic acid 97%
1-Methyl-3-oxocyclobutane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₈O₃
MW: 128.13 g/mol
MDL Number: MFCD00604210
CAS Number: 286442-89-5
UN: 3261
ADR: 8 Ⅲ
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR