(R)-4-Hydroxy-N,N-diphenylpent-2-ynamide 95%

Supplier: Ambeed
Warning

A182743-100MG A182743-1G
77151-648EA 37.08 USD
77151-648 77151-650
(R)-4-Hydroxy-N,N-diphenylpent-2-ynamide 95%
(R)-4-Hydroxy-N,N-diphenylpent-2-ynamide

Precaución: for Research Use Only

Formula: C₁₇H₁₅NO₂
MW: 265.31 g/mol
MDL Number: MFCD28972389
CAS Number: 899809-61-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR