Potassium-4-(hydroxymethyl)phenyltrifluoroborate 98+%

Supplier: Ambeed
Warning

Synonyms: Potassium trifluoro(4-(hydroxymethyl)phenyl)borate

A182570-1G A182570-25G A182570-5G
77151-642EA 15.45 USD
77151-642 77151-644 77151-646
Potassium-4-(hydroxymethyl)phenyltrifluoroborate 98+%
Potassium 4-(hydroxymethyl)phenyltrifluoroborate

Precaución: for Research Use Only

Formula: C₇H₇BF₃KO
MW: 214.04 g/mol
MDL Number: MFCD08276800
CAS Number: 1015082-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR