Ethyl-3-(benzylamino)-3-oxopropionate 97%

Supplier: Ambeed
Warning

A182488-1G A182488-250MG A182488-5G
77151-640EA 30 USD
77151-640 77892-194 77892-195
Ethyl-3-(benzylamino)-3-oxopropionate 97%
Ethyl-3-(benzylamino)-3-oxopropionate

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₃
MDL Number: MFCD28128094
CAS Number: 29689-63-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR