1-Chloro-3,4-dibromo-5-fluorobenzene 95%

Supplier: Ambeed
Warning

A179826-100G A179826-10G A179826-25G A179826-5G
77151-574EA 234 USD
77151-574 77151-576 77151-578 77151-580
1-Chloro-3,4-dibromo-5-fluorobenzene 95%
1-Chloro-3,4-dibromo-5-fluorobenzene

Precaución: for Research Use Only

Formula: C₆H₂Br₂ClF
MDL Number: MFCD00143441
CAS Number: 208186-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR