Lithium 5-methyl-1,3,4-thiadiazole-2-carboxylate 97%

Supplier: Ambeed
Warning

A179754-100MG A179754-250MG
77151-570EA 86 USD
77151-570 77151-572
Lithium 5-methyl-1,3,4-thiadiazole-2-carboxylate 97%
Lithium 5-methyl-1,3,4-thiadiazole-2-carboxylate

Precaución: for Research Use Only

Formula: C₄H₃LiN₂O₂S
MW: 150.08 g/mol
MDL Number: MFCD28975353
CAS Number: 1810070-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR