Methyl 2,5-difluoronicotinate 95%

Supplier: Ambeed
Warning

A178931-1G A178931-250MG
77151-564EA 287.37 USD
77151-564 77151-566
Methyl 2,5-difluoronicotinate 95%
Methyl 2,5-difluoronicotinate

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MW: 173.12 g/mol
MDL Number: MFCD14698038
CAS Number: 1214362-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR