Azetidin-2-ylmethanol 95%

Supplier: Ambeed
Warning

A177185-100MG A177185-1G A177185-250MG
77151-512EA 83.43 USD
77151-512 77151-514 77151-516
Azetidin-2-ylmethanol 95%
Azetidin-2-ylmethanol

Precaución: for Research Use Only

Formula: C₄H₉NO
MW: 87.12 g/mol
MDL Number: MFCD12406977
CAS Number: 250274-91-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR