Purvalanol B 98%

Supplier: Ambeed
Warning

Synonyms: NG95, (R)-2-Chloro-4-((2-((1-hydroxy-3-methylbutan-2-yl)amino)-9-isopropyl-9H-purin-6-yl)amino)benzoic acid

A176253-10MG A176253-1MG A176253-2MG A176253-50MG A176253-5MG
77151-494EA 55 USD
77151-494 77151-496 77151-498 77151-500 77151-502
Purvalanol B 98%
Purvalanol B

Precaución: for Research Use Only

Formula: C₂₀H₂₅ClN₆O₃
MW: 432.9 g/mol
CAS Number: 212844-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR