4-Fluoro-3,5-dimethylbenzyl bromide 95%

Supplier: Ambeed
Danger

A175949-1G A175949-250MG A175949-5G A175949-25G
77151-482EA 45 USD
77151-482 77151-484 77151-486 77901-297
4-Fluoro-3,5-dimethylbenzyl bromide 95%
4-Fluoro-3,5-dimethylbenzyl bromide

Precaución: for Research Use Only

Formula: C₉H₁₀BrF
MDL Number: MFCD06660268
CAS Number: 886501-82-2
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR