1-(3-Bromo-4-nitrophenyl)ethanone 95%

Supplier: Ambeed
Warning

A175680-1G A175680-250MG A175680-5G A175680-10G
77151-470EA 27 USD
77151-470 77151-472 77151-474 77901-550
1-(3-Bromo-4-nitrophenyl)ethanone 95%
1-(3-Bromo-4-nitrophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆BrNO₃
MDL Number: MFCD18973825
CAS Number: 56759-33-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR