6-Iodo-2-oxaspiro[3.3]heptane 97%

Supplier: Ambeed
Warning

A175054-100MG A175054-250MG A175054-1G
77151-466EA 109 USD
77151-466 77151-468 77892-155
6-Iodo-2-oxaspiro[3.3]heptane 97%
6-Iodo-2-oxaspiro[3.3]heptane

Precaución: for Research Use Only

Formula: C₆H₉IO
MDL Number: MFCD22419348
CAS Number: 1363381-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR