4-Bromo-9H-fluoren-9-one 98%

Supplier: Ambeed
Warning

A174942-100MG A174942-1G A174942-250MG A174942-25G A174942-5G
77151-456EA 6 USD
77151-456 77151-458 77151-460 77151-462 77151-464
4-Bromo-9H-fluoren-9-one 98%
4-Bromo-9H-fluoren-9-one

Precaución: for Research Use Only

Formula: C₁₃H₇BrO
MDL Number: MFCD22988837
CAS Number: 4269-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR