H-p-Fluoro-Phe-OEt·HCl 97%

Supplier: Ambeed
Warning

Synonyms: (S)-Ethyl 2-amino-3-(4-fluorophenyl)propanoate hydrochloride, H-p-F-Phe-OEt·HCl, H-p-Fluoro-Phe-OEt·HCl

A174909-1G A174909-250MG A174909-5G
77151-450EA 39.14 USD
77151-450 77151-452 77151-454
H-p-Fluoro-Phe-OEt·HCl 97%
H-p-Fluoro-Phe-OEt·HCl

Precaución: for Research Use Only

Formula: C₁₁H₁₅ClFNO₂
MW: 247.694 g/mol
MDL Number: MFCD02259530
CAS Number: 1534-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR