1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose 98%

Supplier: Ambeed
Warning

A174429-100MG A174429-1G A174429-5G
77151-436EA 15 USD
77151-436 77151-438 77151-440
1,2,3,4-Tetra-O-acetyl-β-D-glucopyranose 98%
1,2,3,4-Tetra-O-acetyl-beta-D-glucopyranose

Precaución: for Research Use Only

Formula: C₁₄H₂₀O₁₀
MDL Number: MFCD00063260
CAS Number: 13100-46-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR