GSK-3β inhibitor VIII 98%

Supplier: Ambeed
Warning

Synonyms: AR-A014418, 1-(4-Methoxybenzyl)-3-(5-nitrothiazol-2-yl)urea, N-(4-Methoxybenzyl)-N′-(5-nitro-1,3-thiazol-2-yl)urea

A174426-100MG A174426-10MG A174426-250MG A174426-25MG A174426-50MG A174426-5MG
77151-422EA 41 USD
77151-422 77151-424 77151-426 77151-428 77151-430 77151-432 77151-434
GSK-3β inhibitor VIII 98%
GSK-3beta inhibitor VIII

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₄O₄S
MDL Number: MFCD08277040
CAS Number: 487021-52-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR