5-Bromo-α,α-dimethyl-2-thiazolemethanol 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-a,a-dimethyl-2-thiazolemethanol

A174290-100MG A174290-1G A174290-250MG
77151-410EA 106.09 USD
77151-410 77151-412 77151-414
5-Bromo-α,α-dimethyl-2-thiazolemethanol 95%
5-Bromo-alpha,alpha-dimethyl-2-thiazolemethanol

Precaución: for Research Use Only

Formula: C₆H₈BrNOS
MW: 222.11 g/mol
MDL Number: MFCD18325135
CAS Number: 879488-37-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR