AZ-3146 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
AZ-3146, AZ 3146, 9-Cyclopentyl-2-[[2-methoxy-4-[(1-methylpiperidin-4-yl)oxy]-phenyl]amino]-7-methyl-7,9-dihydro-8H-purin-8-one, 9-cyclopentyl-2-[2-methoxy-4-(1-methylpiperidin-4-yl)oxyanilino]-7-methylpurin-8-one, 9-cyclopentyl-7,9-dihydro-2-((2-methoxy-4-((1-methyl-4-piperidinyl)oxy)phenyl)amino)-7-methyl-8h-purin-8-one, AZ3146, 9-Cyclopentyl-2-((2-methoxy-4-((1-methylpiperidin-4-yl)oxy)phenyl)amino)-7-methyl-7H-purin-8(9H)-one
A174095-10MG
A174095-1MG
A174095-50MG
A174095-5MG
77151-402EA
12.36
USD
77151-402
77151-404
77151-406
77151-408
AZ-3146 98+%
AZ-3146
Precaución: for Research Use Only
Formula:
C₂₄H₃₂N₆O₃ MW: 452.55 g/mol |
MDL Number:
MFCD18384959 CAS Number: 1124329-14-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...