[RuCl(p-cymene)((S)-binap)]Cl 98%

Supplier: Ambeed
Warning

Synonyms: (S)-RuCl[(p-cymene(BINAP)]Cl, Chloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) chloride [RuCl(p-cymene)((S)-binap)]Cl, Chloro[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl](p-cymene)ruthenium(II) Chloride, [(S)-Binap RuCl p-cymene]Cl

A174018-250MG A174018-25G A174018-5G
77151-392EA 20.6 USD
77151-392 77151-394 77151-396
[RuCl(p-cymene)((S)-binap)]Cl 98%
[RuCl(p-cymene)((S)-binap)]Cl

Precaución: for Research Use Only

Formula: C₅₄H₄₆Cl₄P₂Ru
MW: 999.77 g/mol
MDL Number: MFCD00134456
CAS Number: 130004-33-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR