6,8-Difluoro-3,4-dihydronaphthalen-1(2H)-one 97%

Supplier: Ambeed
Warning

A173608-100MG A173608-250MG A173608-1G
77151-388EA 13 USD
77151-388 77151-390 77567-556
6,8-Difluoro-3,4-dihydronaphthalen-1(2H)-one 97%
6,8-Difluoro-3,4-dihydronaphthalen-1(2H)-one

Precaución: For research use only.

Formula: C₁₀H₈F₂O
MDL Number: MFCD11518781
CAS Number: 895534-38-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR