(S)-Benzyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride 97%

Supplier: Ambeed
Warning

A173124-1G A173124-10G A173124-25G
77151-372EA 45 USD
77151-372 77901-582 77901-448
(S)-Benzyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride 97%
(S)-Benzyl 2-amino-3-(3-(methylsulfonyl)phenyl)propanoate hydrochloride

Precaución: for Research Use Only

Formula: C₁₇H₂₀ClNO₄S
MDL Number: MFCD29919305
CAS Number: 1194550-59-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR