Methyl 3-((tert-butoxycarbonyl)(methyl)amino)propanoate 95%

Supplier: Ambeed
Warning

A173087-100MG A173087-1G A173087-250MG A173087-5G
77151-364EA 11 USD
77151-364 77151-366 77151-368 77151-370
Methyl 3-((tert-butoxycarbonyl)(methyl)amino)propanoate 95%
Methyl 3-((tert-butoxycarbonyl)(methyl)amino)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₉NO₄
MDL Number: MFCD12797747
CAS Number: 119740-95-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR