N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-4',4''-diphenyltriphenylamine

A172932-100MG A172932-1G A172932-250MG A172932-5G
77151-352EA 5.15 USD
77151-352 77151-354 77151-356 77151-358
N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine 95%
N,N-Bis(4-biphenylyl)-N-(4-bromophenyl)amine

Precaución: for Research Use Only

Formula: C₃₀H₂₂BrN
MW: 476.42 g/mol
MDL Number: MFCD20261458
CAS Number: 499128-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR