2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid 95%

Supplier: Ambeed
Danger

Synonyms: 3-Borono-4-fluorobenzohydrazide, 2-Fluoro-5-(hydrazinecarbonyl)phenylboronic acid

A172740-1G A172740-250MG A172740-5G
77151-338EA 173 USD
77151-338 77151-340 77151-342
2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid 95%
2-Fluoro-5-(hydrazinocarbonyl)benzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFN₂O₃
MDL Number: MFCD08436072
CAS Number: 874289-56-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR