Icaritin 98%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 3,5,7-Trihydroxy-2-(4-methoxyphenyl)-8-(3-methylbut-2-en-1-yl)-4H-chromen-4-one, 3,5,7-Trihydroxy-2-(4-methoxyphenyl)-8-(3-methyl-2-buten-1-yl)-4H-1-benzopyran-4-one

A172681-10MG A172681-5MG A172681-100MG A172681-1G A172681-1MG A172681-250MG A172681-5G
77151-334EA 9 USD
77151-334 77151-336 77611-678 77611-680 77611-682 77683-394 77683-395
Icaritin 98%, Ambeed.Inc
Icaritin

Precaución: For research use only.

Formula: C₂₁H₂₀O₆
MW: 368.39 g/mol
MDL Number: MFCD22422519
CAS Number: 118525-40-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR