VE-822 99%

Supplier: Ambeed
Warning

Synonyms: Berzosertib, 5-(4-isopropylsulphonylphenyl)-3-[3-[4-(methylaminomethyl)phenyl]isoxazol-5-yl]pyrazin-2-amine, 5-(4-(isopropylsulphonyl)phenyl)-3-(3-(4-((methylamino)methyl)phenyl)isoxazol-5-yl)pyrazin-2-amine, 3-[3-[4-[(methylamino)methyl]phenyl]-5-isoxazolyl]-5-[4-[(1-methylethyl)sulphonyl]phenyl]- 2-pyrazinamine

A172242-100MG A172242-1MG
77151-314EA 335.78 USD
77151-314 77151-316
VE-822 99%
VE-822

Precaución: for Research Use Only

Formula: C₂₄H₂₅N₅O₃S
MW: 463.55 g/mol
MDL Number: MFCD27976794
CAS Number: 1232416-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR