3-Bromo-4-methoxy-5-nitro-benzoic acid methyl ester 97%

Supplier: Ambeed
Warning

A172143-100MG A172143-1G A172143-250MG A172143-5G
77151-296EA 8 USD
77151-296 77151-298 77151-300 77151-302
3-Bromo-4-methoxy-5-nitro-benzoic acid methyl ester 97%
3-Bromo-4-methoxy-5-nitro-benzoic acid methyl ester

Precaución: for Research Use Only

Formula: C₉H₈BrNO₅
MDL Number: MFCD09881752
CAS Number: 40258-73-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR