2,3-Difluorobenzyl bromide 98%

Supplier: Ambeed
Danger

Synonyms: α-Bromo-2,3-difluorotoluene

A171262-1G A171262-5G A171262-25G
77151-282EA 23 USD
77151-282 77550-104 77567-522
2,3-Difluorobenzyl bromide 98%
2,3-Difluorobenzyl bromide

Precaución: For research use only.

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Boiling Pt: 186
Melting Pt: 8
MDL Number: MFCD00042488
CAS Number: 113211-94-2
UN: 3265
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR